Learn More
Cayman Chemical PIperacetazIn 10mg
SDP
Catalog No. 502910903
Encompass_Preferred
A typical antipsychotic; inhibits spontaneous motor activity and responding for food in rats, as well as potentiates hexobarbital-induced narcosis in mice; inhibits lysosome acidification, as well as viral entry (IC50 = <8.5 µM), in Ebola-infected Huh7 cells
Catalog No. 50-291-0903 Supplier Cayman Chemical Supplier No. 3522010 MG
A typical antipsychotic; inhibits spontaneous motor activity and responding for food in rats, as well as potentiates hexobarbital-induced narcosis in mice; inhibits lysosome acidification, as well as viral entry (IC50 = <8.5 µM), in Ebola-infected Huh7 cells