Learn More
Cayman Chemical PIperacetazIn 50mg
SDP
Catalog No. 502912516
Encompass_Preferred
A typical antipsychotic; inhibits spontaneous motor activity and responding for food in rats, as well as potentiates hexobarbital-induced narcosis in mice; inhibits lysosome acidification, as well as viral entry (IC50 = <8.5 µM), in Ebola-infected Huh7 cells
Catalog No. 50-291-2516 Supplier Cayman Chemical Supplier No. 3522050 MG
A typical antipsychotic; inhibits spontaneous motor activity and responding for food in rats, as well as potentiates hexobarbital-induced narcosis in mice; inhibits lysosome acidification, as well as viral entry (IC50 = <8.5 µM), in Ebola-infected Huh7 cells