Learn More
Medchemexpress LLC Piperacillin-d5 | 61477-96-1 | 96.0% | 522.59 | C23H22D5N5O7S | 10 MG

Supplier: Medchemexpress LLC HYB1923S10MG
Piperacillin-d5 is a deuterium-labeled (d5) analogue of the β-lactam antibiotic piperacillin supplied as a solid powder for use as an internal standard and tracer in quantitative analyses by NMR, GC-MS, and LC-MS. The d5 labeling enables mass-based discrimination with minimal alteration of chemical properties, making it suitable for pharmacokinetic and bioanalytical studies.
- Deuterium-labeled (d5) internal standard for quantitative analysis.
- Solid powder form for analytical sample preparation.
- Purity approximately 96.0%.
- Molecular weight 522.59 g/mol; formula C23H22D5N5O7S.
- Offered in small pack sizes for analytical workflows (for example, 10 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.