Learn More
Medchemexpress LLC Piperacillin-d5 | 61477-96-1 | 98.23% | 1 MG
SDP
Catalog No. 5000412159
Encompass_Preferred
Piperacillin-d5 is deuterium labeled Piperacillin. Piperacillin is a semisynthetic penicillin with a broad spectrum of activity against Gram-positive and Gram-negative aerobic and anaerobic bacteria. It has shown greater activity against β-lactamase-producing organisms than other penicillins.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12159 Supplier Medchemexpress LLC Supplier No. HYB1923S1MG
Piperacillin-d5 is deuterium labeled Piperacillin. Piperacillin is a semisynthetic penicillin with a broad spectrum of activity against Gram-positive and Gram-negative aerobic and anaerobic bacteria. It has shown greater activity against β-lactamase-producing organisms than other penicillins.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.