Learn More
Medchemexpress LLC Piperacillin-d5 | 61477-96-1 | 98.23% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1923S5MG
Encompass_Preferred
Piperacillin-d5 is a deuterium-labeled version of Piperacillin. Piperacillin is a semisynthetic penicillin with a broad spectrum of activity against Gram-positive and Gram-negative aerobic and anaerobic bacteria. It demonstrates greater activity against β-lactamase-producing organisms compared to other penicillins.
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable isotope-labeled compound
  • For research use only
  • Broad spectrum of activity
  • Effective against β-lactamase-producing organisms
Catalog No. 50-004-11243