Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperacillin, G-Biosciences
SCI_ED

Catalog No. S139726
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139726 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139726 Supplier G-Biosciences Supplier No. RC1088
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Piperacillin is an extended-spectrum Beta-lactam antibiotic resistant to many Beta-lactamases. It is derived from ampicillin and targets gram-negative and gram-positive bacteria.

Piperacillin has been used in studies on optimization of antimicrobial therapy in patients with sepsis and to study piperacillin allergic reactions. It has also been used to study multidrug resistant organisms.

Chemical Identifiers

Molecular Weight (g/mol) 517.55

Specifications

Physical Form Powder
Quantity 1 g
Product Line Gibco
Percent Purity 0.98
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.