Learn More
Medchemexpress LLC Piperacillin sodium | 59703-84-3 | 97.9% | 250 MG
SDP
Catalog No. 5000414226
Encompass_Preferred
Piperacillin sodium is a semisynthetic broad-spectrum β-lactam antibiotic which exhibits potent bactericidal activity against Gram-negative bacteria as well as select Gram-positive strains through penicillin-binding proteins. It is most commonly used in combination with the β-lactamase inhibitor Tazobactam.
  • Semisynthetic broad-spectrum β-lactam antibiotic.
  • Exhibits potent bactericidal activity against Gram-negative bacteria and select Gram-positive strains.
  • Acts through penicillin-binding proteins.
  • Most commonly used in combination with the β-lactamase inhibitor Tazobactam.
Catalog No. 50-004-14226 Supplier Medchemexpress LLC Supplier No. 250MGHYB1286R
Piperacillin sodium is a semisynthetic broad-spectrum β-lactam antibiotic which exhibits potent bactericidal activity against Gram-negative bacteria as well as select Gram-positive strains through penicillin-binding proteins. It is most commonly used in combination with the β-lactamase inhibitor Tazobactam.
  • Semisynthetic broad-spectrum β-lactam antibiotic.
  • Exhibits potent bactericidal activity against Gram-negative bacteria and select Gram-positive strains.
  • Acts through penicillin-binding proteins.
  • Most commonly used in combination with the β-lactamase inhibitor Tazobactam.