Learn More
Alkali Scientific Piperacillin, Sodium Salt, 10 G
SDP
Catalog No. 5000661266
Encompass_Preferred
Piperacillin Sodium Salt is a broad-spectrum antibiotic used to treat bacterial infections. It is provided in a powder form with a weight of 10g per bottle. Piperacillin works by inhibiting bacterial cell wall synthesis, preventing bacterial growth and replication. It is commonly used in the treatment of infections caused by both Gram-positive and Gram-negative bacteria. Piperacillin is frequently employed in combination with beta-lactamase inhibitors to extend its efficacy against resistant bacteria. It is utilized in clinical microbiology laboratories for susceptibility testing and is a vital component of antibiotic therapy for severe infections such as sepsis and respiratory tract infections.
Catalog No. 50-006-61266 Supplier Alkali Scientific Supplier No. PIP002CP
Piperacillin Sodium Salt is a broad-spectrum antibiotic used to treat bacterial infections. It is provided in a powder form with a weight of 10g per bottle. Piperacillin works by inhibiting bacterial cell wall synthesis, preventing bacterial growth and replication. It is commonly used in the treatment of infections caused by both Gram-positive and Gram-negative bacteria. Piperacillin is frequently employed in combination with beta-lactamase inhibitors to extend its efficacy against resistant bacteria. It is utilized in clinical microbiology laboratories for susceptibility testing and is a vital component of antibiotic therapy for severe infections such as sepsis and respiratory tract infections.