Learn More
Alkali Scientific Piperacillin, Sodium Salt, 5 G
SDP
Catalog No. 5000661190
Encompass_Preferred
Piperacillin Sodium Salt (5g) is an antibiotic used for the treatment of a variety of bacterial infections. Available in powder form, it is a beta-lactam antibiotic that targets bacterial cell wall synthesis, preventing bacteria from growing and dividing. It is effective against a wide range of both Gram-positive and Gram-negative bacteria. Commonly used in clinical settings, it is typically employed for infections involving the respiratory tract, urinary tract, and intra-abdominal region. This antibiotic is also utilized in susceptibility testing to determine bacterial resistance patterns in microbiology laboratories and is often used in combination with beta-lactamase inhibitors for enhanced efficacy.
Catalog No. 50-006-61190 Supplier Alkali Scientific Supplier No. PIP003CP
Piperacillin Sodium Salt (5g) is an antibiotic used for the treatment of a variety of bacterial infections. Available in powder form, it is a beta-lactam antibiotic that targets bacterial cell wall synthesis, preventing bacteria from growing and dividing. It is effective against a wide range of both Gram-positive and Gram-negative bacteria. Commonly used in clinical settings, it is typically employed for infections involving the respiratory tract, urinary tract, and intra-abdominal region. This antibiotic is also utilized in susceptibility testing to determine bacterial resistance patterns in microbiology laboratories and is often used in combination with beta-lactamase inhibitors for enhanced efficacy.