Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperacillin sodium salt, MilliporeSigma™ Supelco™

Catalog No. 111014810 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4810 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4810 Supplier MilliporeSigma Supelco Supplier No. 93129100MG
Add to Cart
Add to Cart

Piperacillin sodium is a semi-synthetic penicillin which belongs to the β-lactam class of antibiotics with a broad spectrum of antibacterial activity against anaerobic pathogens and both gram positive and gram negative bacteria.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 59703-84-3
Physical Form Neat
Linear Formula C23H26N5 NaO7 S
MDL Number MFCD00917471
Type Analytical Standard
Quantity 100 mg
Formula Weight 539.54
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.