Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperacillin Sodium Salt, MP Biomedicals™

Catalog No. ICN15626801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15626801 1 g
ICN15626805 5 g
ICN15626810 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15626801 Supplier MP Biomedicals, Inc Supplier No. 0215626801
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59703-84-3
Molecular Formula C23H26N5NaO7S
Molecular Weight (g/mol) 539.54
MDL Number MFCD00917471
InChI Key WCMIIGXFCMNQDS-QMJSIVKPSA-M
Synonym Sodium Piperacillin, Pipracil
PubChem CID 23666879
ChEBI CHEBI:8233
SMILES [Na+].CCN1CCN(C(=O)N[C@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O

Specifications

Type Piperacillin Sodium Salt
Physical Form Crystalline Powder
Color White
Quantity 1 g
IUPAC Name sodium;(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Formula Weight 539.5
Chemical Name or Material Piperacillin Sodium Salt
Recommended Storage Store at 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.