Learn More
AdipoGen Piperafizine B
SDP
Supplier:  AdipoGen BVT0446M001
Encompass_Preferred
Chemical. CAS 74720-33-5. Formula C18H14N2O2. MW 290.3. Isolated from Streptomyces thioluteus. Diketopiperazine derivative. Analog of Piperafizine A. Potentiator of vincristine cytotoxicity.
Catalog No. 50-256-1918