Learn More
Cayman Chemical PIperaquIn 100mg
SDP
Catalog No. 502983539
Encompass_Preferred
An antimalarial agent; a bis-quinoline compound; inhibits the ex vivo growth of P. falciparum isolates from malaria-infected patients (IC50s = 11.8-217.3 nM)
Catalog No. 50-298-3539 Supplier Cayman Chemical Supplier No. 20799100 MG
An antimalarial agent; a bis-quinoline compound; inhibits the ex vivo growth of P. falciparum isolates from malaria-infected patients (IC50s = 11.8-217.3 nM)