Learn More
Medchemexpress LLC Piperaquine phosphate | 85547-56-4 | 98.6% | 100 MG
SDP
Catalog No. 5000412401
Encompass_Preferred
Piperaquine phosphate is a bisquinoline antimalarial agent. It can be utilized in antimalarial research in conjunction with Artemisinin.
  • Acts as a bisquinoline antimalarial agent
  • Used in antimalarial research with artemisinin
  • Reduces parasitemia in mice
  • Extends median survival time in animal models with Plasmodium berghei parasites
Catalog No. 50-004-12401 Supplier Medchemexpress LLC Supplier No. HYB1896A100MG
Piperaquine phosphate is a bisquinoline antimalarial agent. It can be utilized in antimalarial research in conjunction with Artemisinin.
  • Acts as a bisquinoline antimalarial agent
  • Used in antimalarial research with artemisinin
  • Reduces parasitemia in mice
  • Extends median survival time in animal models with Plasmodium berghei parasites