Learn More
Medchemexpress LLC Piperaquine (phosphate) | 85547-56-4 | 98.6% | 25 MG
SDP
Catalog No. 5000412549
Encompass_Preferred
Piperaquine phosphate is a bisquinoline antimalarial agent. It can be used in antimalarial research in combination with Artemisinin.
  • Bisquinoline antimalarial agent.
  • Can be used in antimalarial research in combination with Artemisinin.
  • In vivo, Piperaquine (10-90 mg/kg; a single i.p.) decreases parasitemia in mice.
  • At a dose of 90 mg/kg, it resulted in stable body weights and active mice throughout the study.
  • Purity: 98.62%.
Catalog No. 50-004-12549 Supplier Medchemexpress LLC Supplier No. 25MGHYB1896A
Piperaquine phosphate is a bisquinoline antimalarial agent. It can be used in antimalarial research in combination with Artemisinin.
  • Bisquinoline antimalarial agent.
  • Can be used in antimalarial research in combination with Artemisinin.
  • In vivo, Piperaquine (10-90 mg/kg; a single i.p.) decreases parasitemia in mice.
  • At a dose of 90 mg/kg, it resulted in stable body weights and active mice throughout the study.
  • Purity: 98.62%.