Learn More
Medchemexpress LLC HY-B1896B 5mg Medchemexpress, Piperaquine tetraphosphate tetrahydrate CAS:915967-82-7 Purity:>98%
SDP
Catalog No. 501933029
Encompass_Preferred
Medchemexpress, HY-B1896B 5mg Piperaquine tetraphosphate tetrahydrate CAS:915967-82-7 Piperaquine tetraphosphate tetrahydrate is a potent anti-parasites agent, widely used in combination with other antimalarial agents [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3029 Supplier Medchemexpress LLC Supplier No. HYB1896B5MG
Medchemexpress, HY-B1896B 5mg Piperaquine tetraphosphate tetrahydrate CAS:915967-82-7 Piperaquine tetraphosphate tetrahydrate is a potent anti-parasites agent, widely used in combination with other antimalarial agents [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.