Learn More
Medchemexpress LLC Piperazin-2-one-d6 | 1219803-71-0 | 99.3% | 106.16 | 50 MG
SDP
Catalog No. 5000403949
Encompass_Preferred
Piperazin-2-one-d6 is the deuterium-labeled Piperazin-2-one. Piperazin-2-one is a piperazine ring derivative containing amide and ester carbonyl groups. It serves as an important intermediate for synthesizing various active compounds.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-03949 Supplier Medchemexpress LLC Supplier No. HY20128S50MG
Piperazin-2-one-d6 is the deuterium-labeled Piperazin-2-one. Piperazin-2-one is a piperazine ring derivative containing amide and ester carbonyl groups. It serves as an important intermediate for synthesizing various active compounds.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.