Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Piperazin-2-one-d6 | 1219803-71-0 | MFCD11983578 | 99.3% | 106.16 | C4H2D6N2O | 10 MG
SDP

Catalog No. 5000296814
Encompass_Preferred

Piperazin-2-one-d6 is the deuterium-labeled isotopologue of piperazin-2-one, provided as a white solid for use as an internal standard or tracer in analytical and drug-development workflows. It is suited for quantitative mass spectrometry and NMR applications, supplied in small, precisely measured packages, and accompanied by supporting documentation for safe handling and storage.

  • Deuterium-labeled (d6) for isotope dilution quantitation.
  • High purity suitable for analytical standards.
  • White solid stable under recommended storage conditions.
  • Available in multiple small package sizes including 10 mg.
  • Molecular weight 106.16; molecular formula C4H2D6N2O.
  • Supplied with certificate of analysis and safety data sheet.

Catalog No. 50-002-96814 Supplier Medchemexpress LLC Supplier No. HY20128S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Piperazin-2-one-d6 is the deuterium-labeled isotopologue of piperazin-2-one, provided as a white solid for use as an internal standard or tracer in analytical and drug-development workflows. It is suited for quantitative mass spectrometry and NMR applications, supplied in small, precisely measured packages, and accompanied by supporting documentation for safe handling and storage.

  • Deuterium-labeled (d6) for isotope dilution quantitation.
  • High purity suitable for analytical standards.
  • White solid stable under recommended storage conditions.
  • Available in multiple small package sizes including 10 mg.
  • Molecular weight 106.16; molecular formula C4H2D6N2O.
  • Supplied with certificate of analysis and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.