Learn More
Cayman Chemical Piperazine Erastin, Purity: ≥98%, CAS: 1538593-71-3, 10 mg
SDP
Catalog No. 502914873
Encompass_Preferred
A ferroptosis inducer; inhibits glutamate release in human CCF-STTG1 astrocytoma cells (IC50 = 0.8 µM), indicating inhibition of the system xc- cystine/glutamate transporter; decreases GSH levels in HT-1080 cells at 10 µM; inhibits the growth of H-RasG12V-overexpressing BJeLR cells (IC50 = 0.3 µM); increases hepatic mRNA expression of the gene encoding Cox-2 and prevents tumor formation in an HT-1080 mouse xenograft model at 60 mg/kg
Catalog No. 50-291-4873 Supplier Cayman Chemical Supplier No. 3693410 MG
A ferroptosis inducer; inhibits glutamate release in human CCF-STTG1 astrocytoma cells (IC50 = 0.8 µM), indicating inhibition of the system xc- cystine/glutamate transporter; decreases GSH levels in HT-1080 cells at 10 µM; inhibits the growth of H-RasG12V-overexpressing BJeLR cells (IC50 = 0.3 µM); increases hepatic mRNA expression of the gene encoding Cox-2 and prevents tumor formation in an HT-1080 mouse xenograft model at 60 mg/kg