Learn More
Medchemexpress LLC Piperazine-1,4-diylbis(ethane-2,1-diyl) (9Z,9'Z,12Z,12'Z)-bis(octadeca-9,12-dienoate) | 3027565-91-6 | 99.0% | 699.10 g·mol⁻¹ | C44H78N2O4 | 10 MG
SDP
Catalog No. 5000285414
Encompass_Preferred
AA3-DLin is an ionizable cationic lipid used to formulate lipid nanoparticles (LNPs) for mRNA delivery. It has a pKa of 5.8 and is supplied with high purity, making it suitable for research into nucleic acid delivery and vaccine formulation.
  • Ionizable cationic lipid with pKa 5.8 to aid endosomal escape.
  • High purity (99.0%), suitable for research applications.
  • Available in small research quantities for formulation studies.
  • Soluble in ethanol at specified concentrations for LNP preparation.
  • Suitable for formulation and evaluation of mRNA and other nucleic acid delivery systems.
Catalog No. 50-002-85414 Supplier Medchemexpress LLC Supplier No. HY15970010MG
AA3-DLin is an ionizable cationic lipid used to formulate lipid nanoparticles (LNPs) for mRNA delivery. It has a pKa of 5.8 and is supplied with high purity, making it suitable for research into nucleic acid delivery and vaccine formulation.
  • Ionizable cationic lipid with pKa 5.8 to aid endosomal escape.
  • High purity (99.0%), suitable for research applications.
  • Available in small research quantities for formulation studies.
  • Soluble in ethanol at specified concentrations for LNP preparation.
  • Suitable for formulation and evaluation of mRNA and other nucleic acid delivery systems.