Learn More
Medchemexpress LLC Piperazine, 1-(diphenylmethyl)-4-methyl- | 82-92-8 | 98.6% | 25 MG
SDP
Catalog No. 5000414028
Encompass_Preferred
Cyclizine, a piperazine-derivative, is a potent and selective histamine H1 receptor antagonist. It can be used for the research of nausea, vomiting, and dizziness.
  • Potent and selective histamine H1 receptor antagonist.
  • Can be used for the research of nausea, vomiting, and dizziness.
  • For research use only.
  • Solid, white to off-white appearance.
Catalog No. 50-004-14028 Supplier Medchemexpress LLC Supplier No. 25MGHYB1397
Cyclizine, a piperazine-derivative, is a potent and selective histamine H1 receptor antagonist. It can be used for the research of nausea, vomiting, and dizziness.
  • Potent and selective histamine H1 receptor antagonist.
  • Can be used for the research of nausea, vomiting, and dizziness.
  • For research use only.
  • Solid, white to off-white appearance.