Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Piperazine

Catalog No. P459075G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P459075G 5 g
P4590725KG 2.5 kg
P45907500G 500 g
P45907100G 100 g
P459071KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. P459075G Supplier Sigma Aldrich Supplier No. P459075G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Piperazine

Specifications

CAS 94-13-3
Melting Point 95°C to 98°C
Molecular Formula C10H12O3
Linear Formula HOC6H4CO2CH2CH2CH3
MDL Number MFCD00002354
Quantity 5 g
Synonym 1,4-Diazacyclohexane; Diethylenediamine
Molecular Weight (g/mol) 180.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.