Learn More
Medchemexpress LLC Piperazine adipate | 142-88-1 | 98.0% | 1 ML
SDP
Catalog No. 5000412344
Encompass_Preferred
Piperazine adipate is a potent broad spectrum anthelmintic effective against many common worm infections in mammals. It rapidly causes mortality in A. galli and H. gallinae within 30 minutes of exposure. The compound works by significantly inhibiting malate oxidation, aldolase activity, and cholinesterase activity in these parasites. Additionally, it shows inhibitory effects on oxaloacetate reduction.
  • Potent broad-spectrum anthelmintic
  • Effective against common worm infections in mammals
  • Induces rapid mortality in A. galli and H. gallinae
  • Inhibits malate oxidation and aldolase activity in parasites
  • Significantly inhibits cholinesterase activity in Ascaridia galli and Heterakis gallinae
  • Demonstrates inhibitory effects on oxaloacetate reduction
Catalog No. 50-004-12344 Supplier Medchemexpress LLC Supplier No. HYB218610MM/1ML
Piperazine adipate is a potent broad spectrum anthelmintic effective against many common worm infections in mammals. It rapidly causes mortality in A. galli and H. gallinae within 30 minutes of exposure. The compound works by significantly inhibiting malate oxidation, aldolase activity, and cholinesterase activity in these parasites. Additionally, it shows inhibitory effects on oxaloacetate reduction.
  • Potent broad-spectrum anthelmintic
  • Effective against common worm infections in mammals
  • Induces rapid mortality in A. galli and H. gallinae
  • Inhibits malate oxidation and aldolase activity in parasites
  • Significantly inhibits cholinesterase activity in Ascaridia galli and Heterakis gallinae
  • Demonstrates inhibitory effects on oxaloacetate reduction