Learn More
Selleck Chemical LLC Piperazine adipate S4443-100mg
SDP
Catalog No. 503110500
Encompass_Preferred
Piperazine adipate is a broad spectrum anthelmintic agent that inhibits malate oxidation in Ascaridia galli and Heterakis gallinae
Catalog No. 50-311-0500 Supplier Selleck Chemical LLC Supplier No. S4443100MG
Piperazine adipate is a broad spectrum anthelmintic agent that inhibits malate oxidation in Ascaridia galli and Heterakis gallinae