Learn More
Medchemexpress LLC Piperazine erastin | 1538593-71-3 | 99.1% | 645.19 | C35H41ClN6O4 | 10 MG
SDP
Catalog No. 5000212320
Encompass_Preferred
Piperazine erastin is a small-molecule analog of erastin that induces an iron-dependent, non-apoptotic form of cell death called ferroptosis. It is used as a research tool to probe ferroptotic mechanisms and to evaluate therapeutic or mechanistic responses in cellular and in vivo studies.
  • Induces ferroptosis in cell-based assays.
  • High purity suitable for research (99.12%).
  • Supplied as solids and as a 10 mM solution in DMSO.
  • Soluble in DMSO at reported concentrations (~33.33 mg/mL).
  • Documented storage recommendations for powder and in-solvent samples.
Catalog No. 50-002-12320 Supplier Medchemexpress LLC Supplier No. HY10088710MG
Add to Cart
Add to Cart
Piperazine erastin is a small-molecule analog of erastin that induces an iron-dependent, non-apoptotic form of cell death called ferroptosis. It is used as a research tool to probe ferroptotic mechanisms and to evaluate therapeutic or mechanistic responses in cellular and in vivo studies.
  • Induces ferroptosis in cell-based assays.
  • High purity suitable for research (99.12%).
  • Supplied as solids and as a 10 mM solution in DMSO.
  • Soluble in DMSO at reported concentrations (~33.33 mg/mL).
  • Documented storage recommendations for powder and in-solvent samples.