Learn More
Medchemexpress LLC Piperazine erastin | 1538593-71-3 | 99.1% | 645.19 | C35H41ClN6O4 | 25 MG
SDP
Catalog No. 5000211316
Encompass_Preferred
Piperazine erastin is a small-molecule ferroptosis inducer and an analog of erastin. It promotes iron-dependent, non-apoptotic cell death in cellular models and is used in mechanistic and cancer research. The compound is supplied as a high-purity solid or as a DMSO solution and is provided with standard quality and safety documentation for laboratory use.
  • Induces iron-dependent ferroptosis in cell models.
  • High purity suitable for research applications.
  • Available as solid and as a solution in DMSO.
  • Stable under recommended low-temperature storage to preserve activity.
  • Provided with COA and SDS for quality and safety information.
Catalog No. 50-002-11316 Supplier Medchemexpress LLC Supplier No. HY10088725MG
Piperazine erastin is a small-molecule ferroptosis inducer and an analog of erastin. It promotes iron-dependent, non-apoptotic cell death in cellular models and is used in mechanistic and cancer research. The compound is supplied as a high-purity solid or as a DMSO solution and is provided with standard quality and safety documentation for laboratory use.
  • Induces iron-dependent ferroptosis in cell models.
  • High purity suitable for research applications.
  • Available as solid and as a solution in DMSO.
  • Stable under recommended low-temperature storage to preserve activity.
  • Provided with COA and SDS for quality and safety information.