Learn More
Medchemexpress LLC Piperazine erastin | 1538593-71-3 | 99.1% | 645.19 g·mol⁻¹ | C35H41ClN6O4 | 1 MG
SDP
Catalog No. 5000212226
Encompass_Preferred
Piperazine erastin is a small-molecule analog of erastin that triggers an iron-dependent form of non-apoptotic cell death known as ferroptosis. It is provided as a research reagent for cellular and biochemical studies investigating ferroptosis mechanisms and cancer biology.
  • Induces iron-dependent ferroptosis in cell-based assays
  • Suitable for studies of ferroptosis pathways and cancer research
  • High purity appropriate for in vitro experiments
  • Offered in small solid and solution quantities for laboratory use
  • Stable when stored according to recommended temperature conditions
Catalog No. 50-002-12226 Supplier Medchemexpress LLC Supplier No. HY1008871MG
Piperazine erastin is a small-molecule analog of erastin that triggers an iron-dependent form of non-apoptotic cell death known as ferroptosis. It is provided as a research reagent for cellular and biochemical studies investigating ferroptosis mechanisms and cancer biology.
  • Induces iron-dependent ferroptosis in cell-based assays
  • Suitable for studies of ferroptosis pathways and cancer research
  • High purity appropriate for in vitro experiments
  • Offered in small solid and solution quantities for laboratory use
  • Stable when stored according to recommended temperature conditions