Learn More
Medchemexpress LLC Piperazine erastin | 1538593-71-3 | 99.1% | 645.19 g/mol | C35H41ClN6O4 | 50 MG
SDP
Catalog No. 5000211794
Encompass_Preferred
Piperazine erastin is an analog of erastin that induces ferroptosis, an iron-dependent form of non-apoptotic cell death. Supplied for research use, it is used to probe ferroptosis mechanisms and to evaluate anticancer strategies in vitro and in vivo.
  • Induces ferroptosis in cellular assays.
  • High purity (99.1%).
  • Improved solubility and metabolic stability compared with erastin.
  • Available as solid and DMSO solution formulations.
  • Suitable for in vitro and in vivo research applications.
  • Stable when stored under recommended low-temperature conditions.
Catalog No. 50-002-11794 Supplier Medchemexpress LLC Supplier No. HY10088750MG
Piperazine erastin is an analog of erastin that induces ferroptosis, an iron-dependent form of non-apoptotic cell death. Supplied for research use, it is used to probe ferroptosis mechanisms and to evaluate anticancer strategies in vitro and in vivo.
  • Induces ferroptosis in cellular assays.
  • High purity (99.1%).
  • Improved solubility and metabolic stability compared with erastin.
  • Available as solid and DMSO solution formulations.
  • Suitable for in vitro and in vivo research applications.
  • Stable when stored under recommended low-temperature conditions.