Learn More
Selleck Chemical LLC Piperazine Erastin-E1063-25MG
SDP
Catalog No. 503137876
Encompass_Preferred
Piperazine Erastin is an analog of Erastin which can induces ferroptosis in cancer cells an iron-dependent form of nonapoptotic cell death
Catalog No. 50-313-7876 Supplier Selleck Chemical LLC Supplier No. E106325MG
Piperazine Erastin is an analog of Erastin which can induces ferroptosis in cancer cells an iron-dependent form of nonapoptotic cell death