Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Piperidin-2-one | 675-20-7 | 99.9% | 99.13 g/mol | C5H9NO | 10 G
SDP

Catalog No. 5000209634
Encompass_Preferred

2-Piperidone (piperidin-2-one; δ-valerolactam) is an endogenous metabolite and a biomarker for CYP2E1 activity. It is supplied as a high-purity research standard for use in analytical, metabolic, and pharmacological studies. CAS number 675-20-7; molecular formula C5H9NO; molecular weight 99.13 g/mol; purity 99.89%.

  • high-purity research-grade material suitable for analytical assays and reference standards.
  • soluble in DMSO and water at high concentrations with ultrasonic aid.
  • provided in multiple pack sizes to support small-scale and bulk experiments.
  • storage recommendations available for long-term stability in solid and solvent forms.

Catalog No. 50-002-09634 Supplier Medchemexpress LLC Supplier No. HYW04219310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Piperidone (piperidin-2-one; δ-valerolactam) is an endogenous metabolite and a biomarker for CYP2E1 activity. It is supplied as a high-purity research standard for use in analytical, metabolic, and pharmacological studies. CAS number 675-20-7; molecular formula C5H9NO; molecular weight 99.13 g/mol; purity 99.89%.

  • high-purity research-grade material suitable for analytical assays and reference standards.
  • soluble in DMSO and water at high concentrations with ultrasonic aid.
  • provided in multiple pack sizes to support small-scale and bulk experiments.
  • storage recommendations available for long-term stability in solid and solvent forms.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.