Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Piperidin-2-one | 675-20-7 | MFCD00006037 | 99.9% | 99.13 | C5H9NO | 100 G
SDP

Catalog No. 5000209632
Encompass_Preferred

2-Piperidone (piperidin-2-one) is a small lactam used as an endogenous metabolite and analytical standard; it is reported as a biomarker for CYP2E1 activity. It is supplied for research use and includes solubility and storage guidance for assay and formulation work.

  • High purity suitable for analytical use.
  • Soluble in DMSO and water at reported concentrations.
  • Available in multiple pack sizes up to 100 g for bulk use.
  • Storage stability specified for pure form and in solvent.
  • Includes formulation examples for in vivo administration.

Catalog No. 50-002-09632 Supplier Medchemexpress LLC Supplier No. HYW042193100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Piperidone (piperidin-2-one) is a small lactam used as an endogenous metabolite and analytical standard; it is reported as a biomarker for CYP2E1 activity. It is supplied for research use and includes solubility and storage guidance for assay and formulation work.

  • High purity suitable for analytical use.
  • Soluble in DMSO and water at reported concentrations.
  • Available in multiple pack sizes up to 100 g for bulk use.
  • Storage stability specified for pure form and in solvent.
  • Includes formulation examples for in vivo administration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.