Learn More
Medchemexpress LLC Piperidin-4-ol-d4 | 1014695-51-2 | 105.17 | 5 MG
SDP
Catalog No. 5000449233
Encompass_Preferred
Piperidin-4-ol-d4 is the deuterium labeled Piperidin-4-ol. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-49233 Supplier Medchemexpress LLC Supplier No. HYW004328S25MG
Piperidin-4-ol-d4 is the deuterium labeled Piperidin-4-ol. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.