Learn More
Medchemexpress LLC Piperidin-4-ol-d5 | 1219799-35-5 | 106.18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW004328S
Encompass_Preferred
Piperidin-4-ol-d5 is a deuterium labeled variant of Piperidin-4-ol. It incorporates stable heavy isotopes of hydrogen, carbon, and other elements. This compound is primarily utilized as a tracer for quantitation during the drug development process, offering unique advantages due to deuteration's potential to influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
  • Deuterium labeled compound for specialized research.
  • Contains stable heavy isotopes for precise quantitation.
  • Effective as a tracer in drug development studies.
  • May impact drug pharmacokinetics and metabolism.
Catalog No. 50-004-47025