Learn More
Medchemexpress LLC Piperidin-4-ol-d9 | 1219799-39-9 | 110.20 | 10 MG
SDP
Catalog No. 5000448904
Encompass_Preferred
Piperidin-4-ol-d9 is the deuterium labeled Piperidin-4-ol. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled Piperidin-4-ol
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-48904 Supplier Medchemexpress LLC Supplier No. HYW004328S110MG
Piperidin-4-ol-d9 is the deuterium labeled Piperidin-4-ol. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled Piperidin-4-ol
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs