Learn More
Medchemexpress LLC Piperidine, 1-[(4,6-dimethyl-5-pyrimidinyl)carbonyl]-4-[(3S)-4-[(1R)-2-methoxy-1-[4-(trifluoromethyl)phenylethyl]-3-methyl-1-piperazinyl]-4-methyl-, (2Z)-2-butenedioate (1:1) | 599179-03-0 | 99.9% | 649.70 | 25 MG
SDP
Catalog No. 5000384336
Encompass_Preferred
Vicriviroc maleate is a potent, selective, oral bioavailable, and CNS penetrated antagonist of CCR5, with a Ki of 2.5 nM. It also inhibits HIV-1 in PBMC cells. This product is for research use only and not sold to patients.
  • Potent and selective CCR5 antagonist
  • Oral bioavailable and CNS penetrating
  • Inhibits HIV-1 in PBMC cells
  • Available as a white to light yellow solid
  • Ships at room temperature in continental US
Catalog No. 50-003-84336 Supplier Medchemexpress LLC Supplier No. 25MGHY17377
Vicriviroc maleate is a potent, selective, oral bioavailable, and CNS penetrated antagonist of CCR5, with a Ki of 2.5 nM. It also inhibits HIV-1 in PBMC cells. This product is for research use only and not sold to patients.
  • Potent and selective CCR5 antagonist
  • Oral bioavailable and CNS penetrating
  • Inhibits HIV-1 in PBMC cells
  • Available as a white to light yellow solid
  • Ships at room temperature in continental US