Learn More
Medchemexpress LLC Piperidine-2-carboxylic acid | 535-75-1 | MFCD00064347 | 99.0% | 129.16 g/mol | C6H11NO2 | 100mg
SDP
Catalog No. 5000269001
Encompass_Preferred
Pipecolic acid (Standard) is the analytical standard of Pipecolic acid This product is intended for research and analytical applications Pipecolic acid is a metabolite of lysine and an important precursor for many useful microbial secondary metabolites Pipecolic acid slows ferroptosis in diabetic retinopathy by inhibiting the YAP-GPX4 signaling pathway In addition Pipecolic acid can be used as a diagnostic marker for pyridoxine-dependent epilepsy[1][2][3]
Catalog No. 50-002-69001 Supplier Medchemexpress LLC Supplier No. HYY0669R100MG
Pipecolic acid (Standard) is the analytical standard of Pipecolic acid This product is intended for research and analytical applications Pipecolic acid is a metabolite of lysine and an important precursor for many useful microbial secondary metabolites Pipecolic acid slows ferroptosis in diabetic retinopathy by inhibiting the YAP-GPX4 signaling pathway In addition Pipecolic acid can be used as a diagnostic marker for pyridoxine-dependent epilepsy[1][2][3]