Learn More
Medchemexpress LLC Piperidine, 4-[2-[(2,4,6-trifluorophenoxy)methyl]phenyl]-, hydrochloride | 1227056-87-2 | 99.5% | 357.80 g·mol⁻¹ | C18H19ClF3NO | 1 ML
SDP
Catalog No. 5000240486
Encompass_Preferred
Ampreloxetine hydrochloride (TD-9855) is a research-grade small molecule inhibitor of the norepinephrine transporter (NET) and serotonin transporter (SERT). It is CNS-penetrant and supplied for in-vitro research in solid form and as a ready-to-use 10 mM solution in DMSO; intended for research use only.
  • Inhibits NET and SERT for neurotransmitter uptake studies.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • Also available as solid quantities for formulation or storage.
  • High reported purity (99.5%).
  • Molecular weight 357.80 g·mol⁻¹ for dose calculations.
  • Intended for research use only.
Catalog No. 50-002-40486 Supplier Medchemexpress LLC Supplier No. HY10712810MM 1ML
Ampreloxetine hydrochloride (TD-9855) is a research-grade small molecule inhibitor of the norepinephrine transporter (NET) and serotonin transporter (SERT). It is CNS-penetrant and supplied for in-vitro research in solid form and as a ready-to-use 10 mM solution in DMSO; intended for research use only.
  • Inhibits NET and SERT for neurotransmitter uptake studies.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • Also available as solid quantities for formulation or storage.
  • High reported purity (99.5%).
  • Molecular weight 357.80 g·mol⁻¹ for dose calculations.
  • Intended for research use only.