Learn More
Medchemexpress LLC Piperidine, 4-[2-[(4-methylphenyl)thio]phenyl]-, hydrobromide | 960151-65-9 | 99.8% | 364.34 | C18H22BrNS | 50 MG
SDP
Catalog No. 5000245382
Encompass_Preferred
Tedatioxetine hydrobromide is a research reagent that acts as a serotonin- and norepinephrine-preferring triple reuptake inhibitor and as an antagonist at 5-HT2A, 5-HT2C, 5-HT3, and α1A-adrenergic receptors. It is supplied as the hydrobromide salt for preclinical pharmacology and medicinal chemistry studies.
  • Acts as a serotonin- and norepinephrine-preferring triple reuptake inhibitor
  • Antagonizes 5-HT2A, 5-HT2C, 5-HT3, and α1A-adrenergic receptors
  • Supplied as hydrobromide salt with high purity (~99.8%)
  • Available in research pack sizes including 50 mg and a 10 mM solution in DMSO
  • Molecular formula C18H22BrNS; molecular weight 364.34
  • Intended for preclinical research use only; not for human use
Catalog No. 50-002-45382 Supplier Medchemexpress LLC Supplier No. HY10175550MG
Tedatioxetine hydrobromide is a research reagent that acts as a serotonin- and norepinephrine-preferring triple reuptake inhibitor and as an antagonist at 5-HT2A, 5-HT2C, 5-HT3, and α1A-adrenergic receptors. It is supplied as the hydrobromide salt for preclinical pharmacology and medicinal chemistry studies.
  • Acts as a serotonin- and norepinephrine-preferring triple reuptake inhibitor
  • Antagonizes 5-HT2A, 5-HT2C, 5-HT3, and α1A-adrenergic receptors
  • Supplied as hydrobromide salt with high purity (~99.8%)
  • Available in research pack sizes including 50 mg and a 10 mM solution in DMSO
  • Molecular formula C18H22BrNS; molecular weight 364.34
  • Intended for preclinical research use only; not for human use