Learn More
Medchemexpress LLC Piperidine, 4-[2-[(4-methylphenyl)thio]phenyl]-, hydrobromide | 960151-65-9 | 99.8% | 364.34 g/mol | C18H22BrNS | 1 ML
SDP
Catalog No. 5000249020
Encompass_Preferred
Tedatioxetine hydrobromide is the hydrobromide salt of tedatioxetine, a serotonin- and norepinephrine-preferring triple reuptake inhibitor that also antagonizes 5-HT2A, 5-HT2C, 5-HT3, and α1A-adrenergic receptors. It is supplied for research use as a solid and as a 10 mM solution in DMSO, intended for in vitro transporter and receptor pharmacology studies.
  • High purity (99.83%) suitable for research applications.
  • Supplied as solid and as a 10 mM solution in DMSO (1 mL).
  • Suitable for transporter and receptor pharmacology assays.
  • Molecular formula C18H22BrNS; molecular weight 364.34 g/mol.
  • Store sealed, away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-49020 Supplier Medchemexpress LLC Supplier No. HY10175510MM 1ML
Tedatioxetine hydrobromide is the hydrobromide salt of tedatioxetine, a serotonin- and norepinephrine-preferring triple reuptake inhibitor that also antagonizes 5-HT2A, 5-HT2C, 5-HT3, and α1A-adrenergic receptors. It is supplied for research use as a solid and as a 10 mM solution in DMSO, intended for in vitro transporter and receptor pharmacology studies.
  • High purity (99.83%) suitable for research applications.
  • Supplied as solid and as a 10 mM solution in DMSO (1 mL).
  • Suitable for transporter and receptor pharmacology assays.
  • Molecular formula C18H22BrNS; molecular weight 364.34 g/mol.
  • Store sealed, away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).