Learn More
Medchemexpress LLC Piperidine-4-carboxamide-d9 | 1219803-20-9 | 98.2% | 137.23 | 1 MG
SDP
Catalog No. 5000454829
Encompass_Preferred
Piperidine-4-carboxamide-d9 is the deuterium labeled Piperidine-4-carboxamide. It is for research use only and not sold to patients. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54829 Supplier Medchemexpress LLC Supplier No. HYW012748S1MG
Piperidine-4-carboxamide-d9 is the deuterium labeled Piperidine-4-carboxamide. It is for research use only and not sold to patients. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS