Learn More
Medchemexpress LLC Piperidine-4-carboxamide-d9 | 1219803-20-9 | 98.2% | 137.23 | 10 MG
SDP
Catalog No. 5000455120
Encompass_Preferred
Piperidine-4-carboxamide-d9 is the deuterium labeled Piperidine-4-carboxamide. It is a stable heavy isotope of hydrogen, carbon, and other elements incorporated into drug molecules, primarily as a tracer for quantitation during drug development. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis
  • Applicable for NMR, GC-MS, or LC-MS analysis
  • Affects pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-55120 Supplier Medchemexpress LLC Supplier No. HYW012748S10MG
Piperidine-4-carboxamide-d9 is the deuterium labeled Piperidine-4-carboxamide. It is a stable heavy isotope of hydrogen, carbon, and other elements incorporated into drug molecules, primarily as a tracer for quantitation during drug development. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis
  • Applicable for NMR, GC-MS, or LC-MS analysis
  • Affects pharmacokinetic and metabolic profiles of drugs