Learn More
Medchemexpress LLC Piperidine-4-carboxamide-d9 | 1219803-20-9 | 98.2% | 137.23 | 5 MG
SDP
Catalog No. 5000454797
Encompass_Preferred
Piperidine-4-carboxamide-d9 is the deuterium labeled Piperidine-4-carboxamide. It is for research use only. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54797 Supplier Medchemexpress LLC Supplier No. HYW012748S5MG
Piperidine-4-carboxamide-d9 is the deuterium labeled Piperidine-4-carboxamide. It is for research use only. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS