Learn More
Medchemexpress LLC Piperidine, 4-[(diphenylmethoxy)methyl]-1-[3-(4-methoxyphenyl)propyl]-, hydrochloride (1:1) | 136647-02-4 | 99.8% | 466.05 | 10 MG
SDP
Catalog No. 5000454485
Encompass_Preferred
UK-78282 is a novel piperidine, described as a potent and selective Kv1.3 blocker with an IC50 of 200 nM. It effectively suppresses human T-lymphocyte activation in vitro and binds to residues at the inner surface of the channel, overlapping with the site of action of verapamil.
  • Potent and selective Kv1.3 blocker.
  • Effectively suppresses human T-lymphocyte activation in vitro.
  • For research use only.
Catalog No. 50-004-54485 Supplier Medchemexpress LLC Supplier No. HYW01372710MG
UK-78282 is a novel piperidine, described as a potent and selective Kv1.3 blocker with an IC50 of 200 nM. It effectively suppresses human T-lymphocyte activation in vitro and binds to residues at the inner surface of the channel, overlapping with the site of action of verapamil.
  • Potent and selective Kv1.3 blocker.
  • Effectively suppresses human T-lymphocyte activation in vitro.
  • For research use only.