Learn More
Medchemexpress LLC Piperidine-4-sulfonic acid | 72450-62-5 | MFCD00055097 | 100.0% | 165.21 | C5H11NO3S | 5 MG
SDP
Catalog No. 5000222175
Encompass_Preferred
Piperidine-4-sulfonic acid is a small-molecule research reagent and a potent GABA receptor agonist that inhibits H-GABA binding with a reported IC50 of 0.034 μM. The compound has molecular formula C5H11NO3S, molecular weight 165.21 g/mol, and is supplied at high purity for laboratory research use.
  • Potent GABA receptor agonist with an IC50 of 0.034 μM for inhibition of H-GABA binding.
  • High purity (99.95%).
  • Supplied in small milligram pack sizes for research use.
  • Includes product datasheet, certificate of analysis, and safety data sheet.
  • Suitable for neuroscience and GABAergic studies.
Catalog No. 50-002-22175 Supplier Medchemexpress LLC Supplier No. HY1391165MG
Piperidine-4-sulfonic acid is a small-molecule research reagent and a potent GABA receptor agonist that inhibits H-GABA binding with a reported IC50 of 0.034 μM. The compound has molecular formula C5H11NO3S, molecular weight 165.21 g/mol, and is supplied at high purity for laboratory research use.
  • Potent GABA receptor agonist with an IC50 of 0.034 μM for inhibition of H-GABA binding.
  • High purity (99.95%).
  • Supplied in small milligram pack sizes for research use.
  • Includes product datasheet, certificate of analysis, and safety data sheet.
  • Suitable for neuroscience and GABAergic studies.