Learn More
Medchemexpress LLC Piperidine-4-sulfonic acid | 72450-62-5 | MFCD00055097 | C5H11NO3S | 10MG
SDP
Catalog No. 5000260567
Encompass_Preferred
Piperidine-4-sulfonic acid is a small-molecule research reagent that acts as a potent GABA receptor agonist used to probe GABAergic binding and function in in vitro neuroscience studies. It is supplied as a high-purity solid with supporting analytical and safety documentation for laboratory research use.
  • Potent GABA receptor agonist with reported IC50 = 0.034 μM for inhibition of H-GABA binding.
  • High purity (99.95%) suitable for analytical and biological assays.
  • Molecular formula C5H11NO3S and molecular weight 165.21 g·mol⁻¹.
  • Intended for in vitro research use only.
  • Provided with RP-HPLC purity profile and safety data sheet.
Catalog No. 50-002-60567 Supplier Medchemexpress LLC Supplier No. HY13911610MG
Piperidine-4-sulfonic acid is a small-molecule research reagent that acts as a potent GABA receptor agonist used to probe GABAergic binding and function in in vitro neuroscience studies. It is supplied as a high-purity solid with supporting analytical and safety documentation for laboratory research use.
  • Potent GABA receptor agonist with reported IC50 = 0.034 μM for inhibition of H-GABA binding.
  • High purity (99.95%) suitable for analytical and biological assays.
  • Molecular formula C5H11NO3S and molecular weight 165.21 g·mol⁻¹.
  • Intended for in vitro research use only.
  • Provided with RP-HPLC purity profile and safety data sheet.