Learn More
Medchemexpress LLC Piperidine-GNE-049-N-Boc | 1936431-36-5 | 98.5% | C30H39F2N7O2 | 10MG
SDP
Catalog No. 5000261428
Encompass_Preferred
This Boc-protected piperidine ligand is supplied as a solid research reagent used as the target-protein binding fragment in PROTAC design for p300/CBP degraders.
  • High reported purity, 98.5%.
  • Molecular formula C30H39F2N7O2 and molar mass ~567.7 g/mol.
  • Available in multiple small-scale pack sizes suitable for synthesis and analytical work.
  • Solid physical state for easy handling and storage.
  • Intended for research use only; not for therapeutic applications.
Catalog No. 50-002-61428 Supplier Medchemexpress LLC Supplier No. HY13459210MG
This Boc-protected piperidine ligand is supplied as a solid research reagent used as the target-protein binding fragment in PROTAC design for p300/CBP degraders.
  • High reported purity, 98.5%.
  • Molecular formula C30H39F2N7O2 and molar mass ~567.7 g/mol.
  • Available in multiple small-scale pack sizes suitable for synthesis and analytical work.
  • Solid physical state for easy handling and storage.
  • Intended for research use only; not for therapeutic applications.