Learn More
TARGETMOL CHEMICALS INC PIPERIDINE-MO-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T124835MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Piperidine-MO-1 is a dopamine receptor modulator with an ED50 of 68 uMol/kg on increase of DOPAC in the rat striatum (WO/2005/121087A1 compound example 2). purity: 97%
Catalog No. 50-258-1449