Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Piperidinium, 1-(1-cycloocten-1-ylmethyl)-4-[[(2,7-dichloro-9H-xanthen-9-yl)carbonyl]amino]-1-ethyl | 353791-85-2 | 95.2% | 655.44 g·mol⁻¹ | C30H37Cl2IN2O2 | 10 MG
SDP

Catalog No. 5000245203
Encompass_Preferred

A potent, selective small-molecule antagonist of chemokine receptors CCR1 and CCR3 used for receptor pharmacology and anti-inflammatory research.

  • Potent CCR1 antagonist: human IC50 0.9 nM; mouse IC50 5.8 nM.
  • Potent human CCR3 antagonist (IC50 0.58 nM); weak mouse CCR3 activity (IC50 460 nM).
  • Purity 95.2%.
  • Molecular weight 655.44 g·mol⁻¹; chemical formula C30H37Cl2IN2O2.
  • CAS 353791-85-2.
  • Available as solid (mg quantities) and as 10 mM solution in DMSO.
  • Storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-45203 Supplier Medchemexpress LLC Supplier No. HY10336010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A potent, selective small-molecule antagonist of chemokine receptors CCR1 and CCR3 used for receptor pharmacology and anti-inflammatory research.

  • Potent CCR1 antagonist: human IC50 0.9 nM; mouse IC50 5.8 nM.
  • Potent human CCR3 antagonist (IC50 0.58 nM); weak mouse CCR3 activity (IC50 460 nM).
  • Purity 95.2%.
  • Molecular weight 655.44 g·mol⁻¹; chemical formula C30H37Cl2IN2O2.
  • CAS 353791-85-2.
  • Available as solid (mg quantities) and as 10 mM solution in DMSO.
  • Storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.