Learn More
Medchemexpress LLC Piperidinium, 1-(1-cycloocten-1-ylmethyl)-4-[(2,7-dichloro-9H-xanthen-9-yl)carbonyl]amino iodide | 353791-85-2 | MFCD09971097 | 95.2% | 655.44 g/mol | C30H37Cl2IN2O2 | 50 MG
SDP
Catalog No. 5000244928
Encompass_Preferred
J-113863 is a small-molecule CCR1 receptor antagonist provided for research use. It is supplied as solid material and as DMSO solutions and is used in vitro to study chemokine receptor signaling and receptor antagonism.
  • Potent CCR1 antagonist (human IC50 0.9 nM; mouse IC50 5.8 nM).
  • Also a potent human CCR3 antagonist (IC50 0.58 nM).
  • Available as solid and 10 mM DMSO solutions for ready-to-use assays.
  • Molecular weight 655.44 g/mol; formula C30H37Cl2IN2O2; CAS 353791-85-2.
  • Typical purity reported 95.19% (data sheet); batch-specific certificate of analysis available.
  • Intended for in vitro pharmacology and receptor signaling research only.
Catalog No. 50-002-44928 Supplier Medchemexpress LLC Supplier No. HY10336050MG
J-113863 is a small-molecule CCR1 receptor antagonist provided for research use. It is supplied as solid material and as DMSO solutions and is used in vitro to study chemokine receptor signaling and receptor antagonism.
  • Potent CCR1 antagonist (human IC50 0.9 nM; mouse IC50 5.8 nM).
  • Also a potent human CCR3 antagonist (IC50 0.58 nM).
  • Available as solid and 10 mM DMSO solutions for ready-to-use assays.
  • Molecular weight 655.44 g/mol; formula C30H37Cl2IN2O2; CAS 353791-85-2.
  • Typical purity reported 95.19% (data sheet); batch-specific certificate of analysis available.
  • Intended for in vitro pharmacology and receptor signaling research only.