Learn More
Medchemexpress LLC Piperidolate | 82-98-4 | 99.3% | 200 MG
SDP
Catalog No. 5000409099
Encompass_Preferred
Piperidolate is an antimuscarinic that inhibits intestinal cramps induced by acetylcholine in rats and dogs. It is intended for research and analytical applications.
  • Inhibits intestinal cramps
  • Targets muscarinic acetylcholine receptors (mAChR)
  • Relevant for neurological and neurodegenerative diseases
  • Useful for GPCR/G protein and neuronal signaling research
Catalog No. 50-004-09099 Supplier Medchemexpress LLC Supplier No. HYB0962A200MG
Piperidolate is an antimuscarinic that inhibits intestinal cramps induced by acetylcholine in rats and dogs. It is intended for research and analytical applications.
  • Inhibits intestinal cramps
  • Targets muscarinic acetylcholine receptors (mAChR)
  • Relevant for neurological and neurodegenerative diseases
  • Useful for GPCR/G protein and neuronal signaling research